• USA Home
  • GBL900002 - Grace Bio-Labs ProPlate® microarray molded upper structure

EMAIL THIS PAGE TO A FRIEND
GBL900002 Sigma

Grace Bio-Labs ProPlate® microarray molded upper structure

size 2 wells, L × W 16.5 mm × 34 mm

Purchase

Properties

Related Categories Cell Culture Supplies, Labware, ProPlate microarray systems More...
sterility   non-sterile
mfr. no.   Grace Bio-Labs, 900002
L × W   16.5 mm × 34 mm
size   2 wells
well volume   350 μL
color   none
matrix active group   Non-treated surface treatment

Description

Legal Information

ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

ProPlate Microarray Hybridization Plates from Grace BioLabs [VIDEO]

Microarray hybridization system that enables active mixing with minimal reagent use during hybridization. Tutorial on the Proplate microarray hybridization system from Grace Bio-Labs.
Keywords: Microarray Analysis, Nucleic acid hybridization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?