• USA Home
  • GBL910004 - Grace Bio-Labs silicone gasket for ProPlate® microarray system

EMAIL THIS PAGE TO A FRIEND
GBL910004 Sigma

Grace Bio-Labs silicone gasket for ProPlate® microarray system

fits 4 wells, size size 16 mm × 16 mm

Purchase

Properties

Related Categories Cell Culture Supplies, Labware, ProPlate microarray systems More...
sterility   non-sterile
mfr. no.   Grace Bio-Labs, 910004
size size   16 mm × 16 mm
size   4 wells
color   none
matrix active group   Non-treated surface treatment

Description

Legal Information

ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?