EMAIL THIS PAGE TO A FRIEND
GENESYN Sigma

Gene Synthesis

Synthetic DNA for all research applications

Synonym: DNA Sequence, DNA Synthesis, Synthetic DNA Sequence, Synthetic DNA

Purchase

Properties

Related Categories Cell Biology, Genomics Services, Synthetic Biology More...

Description

General description

Gene Synthesis is currently only available to our customers in Europe and some areas in Asia. Please visit this page for more information about our gene synthesis services provided in Europe and some areas of Asia by GENEWIZ. All other customers, please join our mailing list in order to get updates on future availability in your region.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?