• USA Home
  • GEUS79500 - ExoProStar S 5000

EMAIL THIS PAGE TO A FRIEND
GEUS79500 Sigma

ExoProStar S 5000

GE Healthcare, US79500, 5000 reactions

Purchase

Properties

Related Categories Molecular Biology, PCR Clean Up Kits, PCR/Amplification More...
usage   5000 reactions
shelf life   Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
mfr. no.   GE Healthcare, US79500
availability   not available in USA, Canada and Puerto Rico
shipped in   dry ice
storage temp.   −20°C

Description

Analysis Note

To view the Certificate of Analysis for this product, please visit www.gelifesciences.com.

Application

The ExoProStar S PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra ExoProStar S improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.

Features and Benefits

• Easy to automate
• Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
• Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
• Scalable for different reaction sizes
• No loss of PCR product
• Complete heat inactivation of the enzymes within 10 min
• Fast 15 min protocol

General description

ExoProStar S 5000 rxn is an Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs in 15 minutes. For maximum flexibility it comes in two separate tubes.

Legal Information

ExoProStar is a trademark of GE Healthcare

illustra is a trademark of GE Healthcare

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles

Related Content

PCR Selection Guide

Sigma-Aldrich offers a wide variety of PCR reagents to meet any experimental needs. Our range of polymerases is customized to meet your End-Point PCR, qPCR, or RT-PCR needs. Our products vary from ro...
Keywords: Polymerase chain reaction, Polymerase chain reaction - quantitative

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?