EMAIL THIS PAGE TO A FRIEND
GLN2 Sigma

Glutamine Assay Kit

Purchase

Description

Application

A colorimetric assay that is quantitative and linear in the range of 0.25-6 mM L-glutamine.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3316 9
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

GLN1 Glutamine and Glutamate Determination Kit

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?