• USA Home
  • GMATEM - Glycol methacrylate (GMA) kits GMA for TEM

EMAIL THIS PAGE TO A FRIEND
GMATEM Sigma

Glycol methacrylate (GMA) kits GMA for TEM

Purchase

Properties

storage temp.   2-8°C

Description

Other Notes

Components included:
H 8633  2-Hydroxyethyl methacrylate (GMA) 100 ml
B 8637  n-Butyl methacrylate 50 ml
B 2030  Benzoyl Peroxide 5 g

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?