• USA Home
  • H0134 - Hydroxylamine-O-sulfonic acid

EMAIL THIS PAGE TO A FRIEND
H0134 Sigma

Hydroxylamine-O-sulfonic acid

~95%

Purchase

Properties

assay   ~95%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3260 8 / PGIII
WGK Germany 
3
RTECS 
NC5427000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

213136 Hydroxylamine-O-sulfonic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?