• USA Home
  • H0148 - Monoclonal Anti-Heat Shock Protein 25 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
H0148 Sigma

Monoclonal Anti-Heat Shock Protein 25 antibody produced in mouse

clone IAP-9, ascites fluid

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Stress, Antibodies to Chaperonins and Heat Shock Proteins,
species reactivity   human, rat, turkey, rabbit, mouse, guinea pig, chicken, frog
application(s)   immunohistochemistry: suitable using frozen sections of chicken gizzard smooth muscle
  indirect immunofluorescence: suitable using formalin-fixed, heat-shocked cultured chick embryo fibroblasts
  microarray: suitable
clone   IAP-9, monoclonal
antibody form   ascites fluid
isotype   IgG1
mol wt   antigen mol wt 25 kDa
contains   15 mM sodium azide
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... HSPB1(3315)
mouse ... Hspb1(15507)
rat ... Hspb1(24471)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

partially purified inhibitor of actin polymerization (IAP) protein from turkey gizzard smooth muscle.

Antigen Background

Heat Shock Protein 25 (HSP 25) is constitutively expressed in smooth muscle. It is also expressed in fibroblasts following heat shock.

Specificity

A major proteolytic fragment of 22 kDa may also be detected. The antibody suppresses the inhibition of actin elongation induced by the 25 kDa IAP. The antibody displays a punctate or fine fibrillary pattern of labeling throughout the cytoplasm in heat-shocked cells and co-localization with mitochondria. Reactivity has been observed with HSP 25 of various organs.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?