• USA Home
  • H0157 - trans-4-Hydroxy-L-proline-7-amido-4-methylcoumarin hydrochloride

EMAIL THIS PAGE TO A FRIEND
H0157 Sigma

trans-4-Hydroxy-L-proline-7-amido-4-methylcoumarin hydrochloride

  • CAS Number 77471-43-3

  • Empirical Formula (Hill Notation) C15H16N2O4 · HCl

  • Molecular Weight 324.76

  •  MDL number MFCD01310881

  •  PubChem Substance ID 329815104

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   GNFFTQPGTOOZEW-IYJPBCIQSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?