• USA Home
  • H0291 - Anti-HOXB7 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
H0291 Sigma

Anti-HOXB7 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-HHO.C1, Anti-HOX2, Anti-HOX2C, Anti-Homeobox 2C, Anti-Homeobox B7, Anti-Hox-2.3

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Common Transcription Factors, Antibodies to Transcription Factors, Epigenetics,
application(s)   immunoprecipitation: 2.5-5 μg using lysates of HEK-293T cells over expressing human HOXB7
  indirect immunofluorescence: 1-2 μg/mL using paraformaldehyde fixed HEK-293T cells over expressing human HOXB7
  western blot: 1-2 μg/mL using extracts of HEK-293T cells over expressing human HOXB7
clone   polyclonal
concentration   ~1.0 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt ~24 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... HOXB7(3217)
mouse ... Hoxb7(15415)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 103-118 of human HOXB7 conjugated to KLH. The corresponding sequence is identical in mouse.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?