• USA Home
  • H0411 - Hepatocyte Growth Factor Receptor (c-Met)/Fc Chimera from mouse

EMAIL THIS PAGE TO A FRIEND
H0411 Sigma

Hepatocyte Growth Factor Receptor (c-Met)/Fc Chimera from mouse

>95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder

Synonym: HGF Receptor (c-Met)

Purchase

Properties

Related Categories Cell Biology, Cell Culture, Cell Signaling and Neuroscience, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones,
recombinant   expressed in baculovirus infected Sf21 cells
assay   >95% (SDS-PAGE)
form   lyophilized powder
mol wt   calculated mol wt 128 kDa
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   mouse ... Met(17295)

Description

Analysis Note

The biological activity is measured by its ability to bind human HGF.

Biochem/physiol Actions

Heterodimeric transmembrane glycoprotein with binding capacity for c-Met. Activation of this receptor has been correlated with induction of the Ras pathway.

Other Notes

Extracellular domain fused to the C-terminal 6X histidine-tagged Fc region of human IgG1.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

H0540 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H0665 Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse, clone 24516, purified immunoglobulin, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?