• USA Home
  • H0532 - Heparin-Ceramic HyperD M Hydrogel Composite

EMAIL THIS PAGE TO A FRIEND
H0532 Sigma

Heparin-Ceramic HyperD M Hydrogel Composite

saline suspension with ethanol, 80 μm mean particle size

Purchase

Properties

Related Categories Affinity Chromatography, Heparin, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension with ethanol
matrix   Ceramic HyperD M Hydrogel Composite
mean particle size   80 μm
capacity   ≥20 mg/mL binding capacity (antithrombin III)
storage temp.   2-8°C

Description

Features and Benefits

Characteristics are tolerance of very high flow rates without bed compression, and high exchange capacity. Stable to most common solvents, and pH from 2-12.

Other Notes

HyperD media are highly porous, rigid ceramic beads filled with a derivatized hydrogel.

Physical form

Suspension in 1 M NaCl with 20% ethanol

Application

Heparin-Ceramic HyperD M Hydrogel is used in affinity chromatography and protein chromatography. Studies have shown promise for employing heparin-based hydrogels as matrices for in vitro differentiation of hepatocytes or stem cells and as vehicles for transplantation of these cells.

Price and Availability


Detect Your Unseen Protein

Amicon Pro Purification System
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1170C 3 / PGIII
WGK Germany 
1
Flash Point(F) 
73.4 °F
Flash Point(C) 
23 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?