• USA Home
  • H0646 - Anti-HVEM (Herpes virus entry mediator) antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
H0646 Sigma

Anti-HVEM (Herpes virus entry mediator) antibody produced in goat

affinity isolated antibody, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Cytokines and Growth Factors,
species reactivity   human
application(s)   indirect ELISA: 0.5-1 μg/mL
  western blot: 0.1-0.2 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   human ... TNFRSF14(8764)
biological source   goat
conjugate   unconjugated

Description

Immunogen

recombinant human HVEM (herpes virus entry mediator) extracellular domain derived from NS0 cells.

General description

Herpes virus entry mediator (HVEM) also known as ATAR (another TRAF-associated receptor) or TR2 (TNF receptor-like molecule) is a 45kD (approx.) type I membrane protein that belongs to TNF receptor superfamily and is expressed mainly in peripheral blood T cells, B cells, monocytes as well as in tissues enriched in lymphoid cells. During infection it facilitates the herpes virus entry into cell and can inhibit the activation, proliferation and cytokine production of T cells. HVEM interacts with its cellular ligand LIGHT and controls immune response by inducing cell death and cell activation. Anti-HVEM (Herpes virus entry mediator) antibody can be used in indirect ELISA (working concentration:-0.5-1.0 μg/ml). Goat anti-HVEM reacts specifically with recombinant human HVEM. The antibody exhibits no cross-reactivity with recombinant human NGF, TNF sRI and TNF sRII receptor.

Physical form

Lyophilized from 0.2 μm filtered phosphate buffered saline.

Specificity

The antibody does not cross-react with recombinant human TNF sRI, recombinant human TNF sRII, and recombinant human NGF-R.

Application

Anti-HVEM (Herpes virus entry mediator) antibody may be used for immunoblotting at a working antibody concentration of 0.1-0.2 μg/ml for detecting human HVEM.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?