• USA Home
  • H0647 - 5-Hydroxy-N-methyl-2-pyrrolidinone ethyl ether

EMAIL THIS PAGE TO A FRIEND
H0647 Sigma

5-Hydroxy-N-methyl-2-pyrrolidinone ethyl ether

~95% (HPLC)

Synonym: 5-Ethoxy-N-methyl-2-pyrrolidinone

Purchase

Properties

assay   ~95% (HPLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?