EMAIL THIS PAGE TO A FRIEND
H0761 Sigma

Silica gel

30 μm particle size (average), average pore diameter 60 Å, Suitable for normal-phase adsorption-partition chromatography

Purchase

Properties

Related Categories Essential Chemicals, Other Silica Gels, Research Essentials, Silica Gels
InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N
particle size   30 μm (average)
pore size   60 Å average pore diameter
surface area   500 m2/g
bp   2230 °C
mp   >1600 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?