EMAIL THIS PAGE TO A FRIEND
H100 Sigma

Haloperidol

Synonym: 4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone, 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone

Purchase

Properties

InChI Key   LNEPOXFFQSENCJ-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 3
WGK Germany 
3
RTECS 
EU1575000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

H1512 Haloperidol, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?