• USA Home
  • H1010 - N-Heptyl-L-hydroxyproline

EMAIL THIS PAGE TO A FRIEND
H1010 Sigma

N-Heptyl-L-hydroxyproline

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   QWMDYNIEGNYUSY-UHFFFAOYSA-N
storage temp.   −20°C

Description

Application

HPLC stationary phase for resolution of α-amino acids.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?