EMAIL THIS PAGE TO A FRIEND
H1034 Sigma
Hybri-slips
L × W × thickness 40 mm × 24 mm × 0.25 mm
Purchase
Description
Application
Hybri-slips are hydrophobic cover slips for in situ hybridization, in situ PCR, and hybridization to genomic arrays on glass slides.
General description
• Constant probe concentration - Unlike glass cover slips, Hybri-slips do not adsorb probes.
• Ready-to-use - Hybri-slips do not require pre-treatment prior to use.
• Avoid RNA degradation - Hybri-slips are supplied RNAse-free and are protected from contamination by clean-release liners.
• Durable - Hybri-slips remain flat even at elevated temperatures.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?