• USA Home
  • H1282 - halfTERM Dye Terminator Sequencing Reagent

EMAIL THIS PAGE TO A FRIEND
H1282 Sigma

halfTERM Dye Terminator Sequencing Reagent

Purchase

Properties

storage temp.   −20°C

Description

Application

For use with Amersham-Pharmacia Biotech or ABI terminator premixes.

Legal Information

halfTERM is a trademark of Genpak Ltd.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?