• USA Home
  • H1389 - n-Heptyl β-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
H1389 Sigma

n-Heptyl β-D-glucopyranoside

~98% (GC)

  • CAS Number 78617-12-6

  • Empirical Formula (Hill Notation) C13H26O6

  • Molecular Weight 278.34

  •  Beilstein Registry Number 8806

  •  MDL number MFCD00063299

  •  PubChem Substance ID 24895455

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Non-Ionic Detergents
description   non-ionic
InChI Key   NIDYWHLDTIVRJT-UJPOAAIJSA-N
assay   ~98% (GC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?