• USA Home
  • H1391 - 3-Hydroxytetrahydrofuran

EMAIL THIS PAGE TO A FRIEND
H1391 Sigma

3-Hydroxytetrahydrofuran

Purchase

Properties

density   1.09 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
LU3450050
Flash Point(F) 
178 °F
Flash Point(C) 
81 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H59109 3-Hydroxytetrahydrofuran, ReagentPlus®, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?