• USA Home
  • H1392 - Harmaline hydrochloride dihydrate

EMAIL THIS PAGE TO A FRIEND
H1392 Sigma

Harmaline hydrochloride dihydrate

≥95%

Synonym: 1-Methyl-7-methoxy-3,4-dihydro-β-carboline hydrochloride

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neurobiology, Neuroscience, Neurotransmission (Obesity),
InChI Key   LCEKUHFBUFUSSY-UHFFFAOYSA-N
assay   ≥95%
drug control   stupéfiantregulated under CDSA - not available from Sigma-Aldrich Canada
storage temp.   −20°C

Description

Biochem/physiol Actions

CNS stimulant; may act through NMDA receptors

Packaging

1 g in poly bottle

100, 500 mg in poly bottle

Price and Availability


Neuroscience Grant Programm
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MG8750000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?