• USA Home
  • H1394 - (±)-5-Hydroxy-(6E,8Z,11Z,14Z,17Z)-eicosapentaenoic acid

EMAIL THIS PAGE TO A FRIEND
H1394 Sigma

(±)-5-Hydroxy-(6E,8Z,11Z,14Z,17Z)-eicosapentaenoic acid

~100 μg/mL in ethanol, ≥98%

Synonym: (±)-5-HEPE

Purchase

Properties

assay   ≥98%
concentration   ~100 μg/mL in ethanol
storage temp.   −20°C

Description

Packaging

Packaged under Argon

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?