• USA Home
  • H1407 - halfBD Dye Terminator Sequencing Reagent

EMAIL THIS PAGE TO A FRIEND
H1407 Sigma

halfBD Dye Terminator Sequencing Reagent

for DNA sequencing

Purchase

Properties

grade   for molecular biology
form   liquid
storage temp.   −20°C

Description

Application

Suitable for use with the ABI BigDye® terminator premix solution for sequencing.

Features and Benefits

• Reduces the amount of ABI BigDye Terminator Premix required
• No reduction in accuracy, resolution, read length, or signal strength
• Compatable with ABI Prism 3700, 3100 & 310 DNA sequencers
• Also compatable with Molecular Dynamics MegaBACE instruments

General description

Sigma’s halfBD Dye Terminator Sequencing Reagent is specially formulated to give identical resolution, read length, accuracy and signal strength compared to the standard ABI protocol.

Legal Information

BigDye is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries

halfBD is a trademark of Genpak Ltd.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

B4930 Capillary Electrophoresis Running Buffer (10x), for automated DNA sequencing
S3938 SeqSaver Sequencing Premix Dilution Buffer, for DNA sequencing
92210 Timestrip Plus 0 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?