• USA Home
  • H1409 - Hybri-well Press-seal hybridization chambers
EMAIL THIS PAGE TO A FRIEND
H1409 Sigma
Hybri-well Press-seal hybridization chambers
chamber L × W × D 32 mm × 19 mm × 0.15 mm
Purchase
Description
General description
Hydrophobic cover slip with non-cytotoxic spacer adheres to a microscope slide, forming a chamber with access ports for addition and removal of reagents. RNase-free, heat-resistant, and disposable: ideal for hybridization. Easily removed from microscope slide after hybridization steps.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?