EMAIL THIS PAGE TO A FRIEND
H1513 Sigma

Human Serum

from clotted human male whole blood, Type AB, sterile-filtered, (mycoplasma tested, virus tested)

Purchase

Properties

Related Categories Cell Culture, Fetal Bovine Serum (FBS) and Other Animal Sera, Human Sera More...
biological source   from clotted human male whole blood
sterility   sterile-filtered
impurities   (mycoplasma tested, virus tested)
shipped in   dry ice
storage temp.   −20°C

Description

Analysis Note

Endotoxin tested

Warning

Each donor is tested for and found non-reactive for Hepatitis B & C and non-reactive for Human Immunodeficiency Virus (HIV) antibody by ELISA. Nevertheless, products of human origin should be considered potentially infectious and handled accordingly.

Price and Availability


Cell Freezing

All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

H4522 Human Serum, from human male AB plasma, USA origin, sterile-filtered

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?