• USA Home
  • H1521 - 6β-Hydroxy-8,13-epoxy-labd-14-en-11-one from Coleus forskohlii

EMAIL THIS PAGE TO A FRIEND
H1521 Sigma

6β-Hydroxy-8,13-epoxy-labd-14-en-11-one from Coleus forskohlii

≥98% (TLC)

Synonym: 6β-Hydroxy-4-deoxycoleol

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cyclic Nucleotide Metabolism, G Proteins and Cyclic Nucleotides, Guanylyl Cyclase Activators More...
InChI Key   QSBSWAUSLLQUDV-FCZHJMSUSA-N
assay   ≥98% (TLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?