EMAIL THIS PAGE TO A FRIEND
H166 Sigma

p-Hydroxycocaine

  • CAS Number 89339-16-2

  • Empirical Formula (Hill Notation) C17H21NO5

  • Molecular Weight 319.35

  • Popular Documents:  Datasheet (PDF)  

Purchase

Properties

form   neat
drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 1544PSN1 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?