• USA Home
  • H1768 - Anti-Horse Serum antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
H1768 Sigma

Anti-Horse Serum antibody produced in rabbit

whole antiserum

Synonym: Anti-Serum, horse

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Animal Plasma Proteins, HJ-HO, Primary Antibodies,
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
contains   15 mM sodium azide
species reactivity   horse
application(s)   immunoelectrophoresis: suitable
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Other Notes

Antiserum adsorbed with bovine and porcine serum proteins

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Search 80,000 Antibodies at Sigma-Aldrich

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?