• USA Home
  • H1876 - Hyaluronic acid sodium salt from human umbilical cord

EMAIL THIS PAGE TO A FRIEND
H1876 Sigma

Hyaluronic acid sodium salt from human umbilical cord

Synonym: Poly(β-glucuronic acid-[1→3]-β-N-acetylglucosamine-[1→4]) alternating, Sodium hyaluronate

Purchase

Properties

Related Categories Carbohydrate, Cell Biology, Metabolic Pathways, Metabolites and Cofactors on the Metabolic Pathways Chart, Metabolomics More...
InChI Key   MAKUBRYLFHZREJ-JWBQXVCJSA-M
impurities   ≤5% Protein (Lowry)
intrinsic viscosity   ≥60 (not determined by Sigma)(lit.)
storage temp.   −20°C

Description

Quality

Also contains chondroitin sulfate.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
MT7250000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?