• USA Home
  • H1897 - Handbook of Analytical Derivatization Reactions
EMAIL THIS PAGE TO A FRIEND
H1897 Sigma
Handbook of Analytical Derivatization Reactions
D. Knapp, John Wiley & Sons, Ltd, 1979, 741 pp., hard cover
  •  ISBN-10 0-471-03469-X
  •  ISBN-13 978-0-471-03469-8
Purchase
Properties
publication info   D. Knapp, John Wiley & Sons, Ltd, 1979, 741 pp., hard cover
mfr. no.   Wiley
Description
General description
Much of the analytical work involving GC, MS, GC-MS and more recently HPLC involve analytical derivatization prior to analysis. This book presents a collection of these derivatization methods. Those described here involve covalent derivatives of organic compounds.
Table of Contents
Introduction: Uses of Analytical Derivatization.  Derivative Types and Reagents.  General Considerations in Analytical Derivatization.  Apparatus for Analytical Derivatization.  Derivatization of Particular Compound Types: Hydroxyl, Sulfhydryl, and Epoxy Compounds.  Amino Compounds.  Carboxylic Acids.  Unsaturated, Cyclopropane, and Methyl Branched Fatty Acids: Derivatives for Determination of Substituent Location.  Amino Acids and Peptides.  Aldehydes and Ketones.  Nitrogen Functional Groups Other than the Amino Group.  Phospho and Sulfo Compounds.  Derivatives for Chromatographic Separation of Optical Isomers.  Fatty Lipids.  Steroids, Prostaglandins and Related Compounds.  Carbohydrates.  Nucleotides, Their Constituents, and Related Compounds.  Drugs.  Appendices.  Indexes.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?