• USA Home
  • H1898 - Helicobacter TISSUE-TROL

EMAIL THIS PAGE TO A FRIEND
H1898 Sigma

Helicobacter TISSUE-TROL

from mouse intestine tissue containing Helicobacter pylori

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, TISSUE-TROL Control Slides More...
Quality Level   GMP-IVD
biological source   from mouse intestine tissue containing Helicobacter pylori
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Components

25 slides per box. 1 stained slide is provided with each box. The stained slide has been stained with Sigma Procedure No. HT101, a modified Steiner-Steiner procedure.

Application

Suitable for use with Sigma Silver Stain, Modified Steiner-Steiner, Procedure No. HT101.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

TTR006 Helicobacter TISSUE-TROL Control Slides, from mouse intestine tissue containing Helicobacter pylori

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?