• USA Home
  • H1909 - Modular hybridization system
EMAIL THIS PAGE TO A FRIEND
H1909 Sigma
Modular hybridization system
Assortment pack
Purchase
Description
Compatibility
Reusable, press-to-seal gasket and cover slip enclosures which can be quickly assembled and disassembled for hybridizing and imaging thick or free floating specimens of any thickness with up to a 13 mm diameter. Build up to three chambers per slide. Minimize specimen handling, eliminate evaporation and leaking even when submerged. RNase-free, ready to use for multiple incubations and agitated washing steps. (Assortment pack contains seal tabs, 5 ea A-D & 10 each E).
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?