• USA Home
  • H2010 - Hemoglobin from baboon plasma

EMAIL THIS PAGE TO A FRIEND
H2010 Sigma

Hemoglobin from baboon plasma

lyophilized powder

Synonym: Hb

Purchase

Properties

Related Categories Biochemicals and Reagents, Hemoglobin, Plasma & Blood Proteins, Plasma, Blood, and Related Proteins and Reagents, Proteins and Derivatives More...
form   lyophilized powder
storage temp.   2-8°C

Description

Biochem/physiol Actions

Oxygen transporter, NO scavenger

Caution

Since native hemoglobin is readily oxidized in air, these preparations may be predominantly methemoglobin.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Cellular Derived Blood Proteins and Enzymes

Cell damage is induced by reactive oxygen species (ROS). ROS are either free radicals, reactive anions containing oxygen atoms, or molecules containing oxygen atoms that can either produce free radic...
BioFiles 2006, 1.5, 4.
Keywords: Aerobic, Alzheimer Disease, Bacterial conjugations, Biofiles, Cancer, Metabolism, Oxidations, Parkinson Disease, Reductions, Respiration

HPLC Analysis of Hemoglobin Variants on Discovery® BIO PolyMA-WAX

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Hemoglobin Variants on Discovery® BIO PolyMA-WAX
Keywords: Chromatography, High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?