• USA Home
  • H2269 - Hydrocodone (+)-bitartrate salt solution

EMAIL THIS PAGE TO A FRIEND
H2269 Sigma

Hydrocodone (+)-bitartrate salt solution

drug standard, 1.0 mg/mL in methanol

  • Empirical Formula (Hill Notation) C18H21NO3 · C4H6O6 · xH2O

  • Molecular Weight 449.45 (anhydrous basis)

  •  EC Number 200-659-6

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Opiates / Synthetic Analgesic Drug Standards, Pharmaceuticals, Illicit Drugs & Alcohol
grade   drug standard
assay   ≥98%
drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?