EMAIL THIS PAGE TO A FRIEND
H2411 Sigma

Tissue slide

from human normal adult, multiple organ

Purchase

Description

Application

Suitable for immunohistochemistry and in situ hybridization. May be used for cellular localization of tissue specific mRNA and protein expression.

General description

Each slide contains sections from 8 Human Adult Normal tissues: Esophagus, Stomach, Small Intestine, Colon, Uterus, Placenta, Bladder, Adipose

Preparation Note

Paraffin embeded tissue sections, approximately 5 μm thick.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?