EMAIL THIS PAGE TO A FRIEND
H2514 Sigma

1-Hexacosyl acetate

Synonym: Acetic acid hexacosyl ester

  • CAS Number 822-32-2

  • Empirical Formula (Hill Notation) C28H56O2

  • Molecular Weight 424.74

  •  MDL number MFCD00057783

  •  PubChem Substance ID 24895504

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?