• USA Home
  • H2645 - 15(S)-Hydroxy-(5Z,8Z,11Z,13E)-eicosatetraenoic acid methyl ester

EMAIL THIS PAGE TO A FRIEND
H2645 Sigma

15(S)-Hydroxy-(5Z,8Z,11Z,13E)-eicosatetraenoic acid methyl ester

~75 μg/mL in methanol

Purchase

Properties

concentration   ~75 μg/mL in methanol
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1230 3/PG 2
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?