• USA Home
  • H2897 - (S)-α-(Hydroxymethyl)tyrosine

EMAIL THIS PAGE TO A FRIEND
H2897 Sigma

(S)-α-(Hydroxymethyl)tyrosine

Synonym: α-Hydroxymethyl-D-tyrosine, (−)-α-(Hydroxymethyl)tyrosine

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   KQAABALABGCGPM-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?