• USA Home
  • H2900 - Hybridoma Enhancing Supplement from murine thymoma cell line

EMAIL THIS PAGE TO A FRIEND
H2900 Sigma

Hybridoma Enhancing Supplement from murine thymoma cell line

liquid, sterile-filtered, suitable for hybridoma

Synonym: Conditioned Medium from murine thymoma cell line

Purchase

Properties

Related Categories Cell Culture, Conditioned Media, Media Supplements and Reagents, Hybridoma Media Supplements, Hybridoma Platform, Hybridoma Reagents,
sterility   sterile-filtered
form   liquid
quality   endotoxin tested, mycoplasma tested, hybridoma tested.
suitability   suitable for hybridoma
shipped in   dry ice
storage temp.   −20°C

Description

Application

Use at a concentration of 10% in complete medium containing FBS (10%) or CPRS-3. Optimal conditions must be determined for specific applications.

Caution

Avoid repeated freezing and thawing. If entire contents are not used, store frozen in working aliquots. Precipitation may occur when stored. This is not detrimental to the product and may be reversed when warmed to 37 °C.

Other Notes

Conditioned medium from a proprietary murine thymoma cell line grown in RPMI-1640 with 15% FBS and IL-6.

Price and Availability


Filter out uncertainty.
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

H6020 Hybridoma Enhancing Supplement Hybri-Max from murine thymoma cell line, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?