EMAIL THIS PAGE TO A FRIEND
H3025 Sigma

Heparin−Agarose

Type II-S, saline suspension, ~170 USP units/mg ligand

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics, Specialty Resins More...
biological source   Heparin from porcine intestinal mucosa
type   Type II-S
form   saline suspension
extent of labeling   400-800 μg per mL
matrix   Cross-linked 4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   amino
matrix spacer   1 atom
capacity   ≥0.5 mg/mL binding capacity (thrombin)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

H6508 Heparin−Agarose, Type I, saline suspension

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?