EMAIL THIS PAGE TO A FRIEND
H3131 Sigma

2-Hydroxyestradiol

≥95% (HPLC)

Synonym: 1,3,5(10)-Estratriene-2,3-17β-triol, Estra-1,3,5(10)-triene-2,3,17-triol, NSC 61711

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones,
InChI Key   DILDHNKDVHLEQB-XSSYPUMDSA-N
assay   ≥95% (HPLC)
storage temp.   −20°C
Gene Information   rat ... Ar(24208)

Description

Packaging

1, 5 mg in glass insert

10 mg in glass bottle

Price and Availability


Better Together

Discover your Target

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KG7675000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?