• USA Home
  • H3260 - 1-O-Palmityl-2,3-dipalmitoyl-rac-glycerol

EMAIL THIS PAGE TO A FRIEND
H3260 Sigma

1-O-Palmityl-2,3-dipalmitoyl-rac-glycerol

~99%

Synonym: 1,2-Dipalmitoyl-3-O-palmityl-rac-glycerol, 1-O-Hexadecyl-2,3-dihexadecanoyl-rac-glycerol

  • CAS Number 1116-45-6

  • Empirical Formula (Hill Notation) C51H100O5

  • Molecular Weight 793.34

  •  MDL number MFCD00048477

  •  PubChem Substance ID 24895564

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Ether Lipid Metabolism, Glycerolipids, Lipids,
InChI Key   PPDBIVVWNXSKCW-UHFFFAOYSA-N
assay   ~99%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?