• USA Home
  • H3266 - L-Histidine Decarboxylase from Lactobacillus 30a

EMAIL THIS PAGE TO A FRIEND
H3266 Sigma

L-Histidine Decarboxylase from Lactobacillus 30a

Type III-S, crude acetone powder, 0.25-0.5 unit/mg solid

Synonym: L-Histidine carboxylyase

Purchase

Description

Application

Major synthetic enzyme for histamine; decarboxylates L-histidine to form histamine

Quality

Activity may be extracted with 0.2 M ammonium acetate, pH 4.8.

Unit Definition

One unit will release 1.0 μmole of CO2 from L-histidine per min at pH 4.5 at 37 °C.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?