EMAIL THIS PAGE TO A FRIEND
H3379 Sigma

Hydantoic acid

Synonym: N-Carbamoylglycine

Purchase

Properties

InChI Key   KZVRXPPUJQRGFN-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
MB7660000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?