• USA Home
  • H3392 - Deaminated heparin−Agarose

EMAIL THIS PAGE TO A FRIEND
H3392 Sigma

Deaminated heparin−Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics, Specialty Resins More...
form   saline suspension
extent of labeling   12-18 mg per mL (by carbazole assay)
matrix   cross-linked 4% beaded agarose
matrix activation   epichlorohydrin
matrix attachment   reductive amination through aldehyde groups of heparin
matrix spacer   7 atoms
capacity   12-18 mg/mL binding capacity (bovine antithrombin III)
storage temp.   2-8°C

Description

Physical form

Suspension in 1.0 M NaCl containing 10% ethanol

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?