• USA Home
  • H4003 - 16α-Hydroxyetiocholanolone

EMAIL THIS PAGE TO A FRIEND
H4003 Sigma

16α-Hydroxyetiocholanolone

≥97% (TLC)

Synonym: 3α,16α-Dihydroxy-5β-androstan-17-one, 5β-Androstane-3α,16α-diol-17-one

Purchase

Properties

InChI Key   HLQYTKUIIJTNHH-NMTBMHBSSA-N
assay   ≥97% (TLC)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?