EMAIL THIS PAGE TO A FRIEND
H4137 Sigma

L-Homocystine

cell culture tested

Synonym: L-4,4′-Dithiobis(2-aminobutanoic acid)

  • CAS Number 626-72-2

  • Empirical Formula (Hill Notation) C8H16N2O4S2

  • Molecular Weight 268.35

  •  Beilstein Registry Number 1728583

  •  EC Number 210-962-5

  •  MDL number MFCD00020391

Purchase

Properties

InChI Key   ZTVZLYBCZNMWCF-WDSKDSINSA-N
mp   281-284 °C (dec.)(lit.)
suitability   cell culture tested

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?