• USA Home
  • H4144 - (±)-9-Hydroxy-(5Z,7E,11Z,14Z)-eicosatetraenoic acid

EMAIL THIS PAGE TO A FRIEND
H4144 Sigma

(±)-9-Hydroxy-(5Z,7E,11Z,14Z)-eicosatetraenoic acid

~100 μg/mL in ethanol, ~98%

Synonym: (±)-9-HETE

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acids and conjugates, Fatty Acyls, Hydroxy fatty acids, Lipids,
InChI Key   KATOYYZUTNAWSA-OIZRIKEUSA-N
assay   ~98%
concentration   ~100 μg/mL in ethanol
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

Packaged under Argon.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?