EMAIL THIS PAGE TO A FRIEND
H4146 Sigma

Hippuryl-Arg-Gly

≥97% (HPLC)

Synonym: N-Benzoyl-Gly-Arg-Gly

  • CAS Number 96723-71-6

  • Empirical Formula (Hill Notation) C17H24N6O5

  • Molecular Weight 392.41

  •  MDL number MFCD00133398

  •  PubChem Substance ID 24895624

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Chromogenic, Enzyme Substrates, Enzymes, Inhibitors, and Substrates More...
InChI Key   QAYUGHWUEAJNPD-UHFFFAOYSA-N
assay   ≥97% (HPLC)
composition   Peptide content, ~70%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?